Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum 30 wt% Rhodium wire, 0.5mm (0.02 in.) dia., ISA Type B Standard Grade Thermocouple

For ISA Type B Standard Grade Thermocouple

Chemical Identifiers

Chemical Name or Material Platinum 30 wt% Rhodium wire
Name Note 0.5mm (0.02 in.) dia., ISA Type B Standard Grade Thermocouple
MDL Number MFCD00198186
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12576BY

  SDS
alfa aesar™
12576BY
100cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12576CC

  SDS
alfa aesar™
12576CC
500cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 70:30 wt%
Chemical Name or Material Platinum 30 wt% Rhodium wire
TSCA TSCA
Physical Form Wire
Quantity 100cm
Weight ≈0.95g/25cm
Odor Odorless
MDL Number MFCD00198186
Name Note 0.5mm (0.02 in.) dia.