Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 3% on carbon paste, B103032-3

Chemical Identifiers

Chemical Name or Material Platinum
CAS 7440-06-4
Name Note 3% on carbon paste, B103032-3
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3630106

  SDS
alfa aesar™
H3630106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3630114

  SDS
alfa aesar™
H3630114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3630122

  SDS
alfa aesar™
H3630122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum
Name Note 3% on carbon paste, B103032-3
CAS 7440-06-4
TSCA TSCA
Physical Form Powder
MDL Number MFCD00011179
Odor Odorless
Color Black
Quantity 5g