Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 3% on activated carbon powder, standard, unreduced, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Platinum
Name Note 3% on activated carbon powder, Standard, unreduced, nominally 50% water wet
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3831706

  SDS
alfa aesar™
3831706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3831714

  SDS
alfa aesar™
3831714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3831722

  SDS
alfa aesar™
3831722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Weight 900 to 1100m2/g
MDL Number MFCD00011179
Chemical Name or Material Platinum
Quantity 5g
Name Note 3% on activated carbon powder, Standard, unreduced, nominally 50% water wet
TSCA TSCA