Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum 13 wt% Rhodium wire, 0.5mm (0.02 in.) dia., ISA Type R Standard Grade Thermocouple

For ISA Type R Standard Grade Thermocouple

Chemical Identifiers

Chemical Name or Material Platinum 13 wt% Rhodium wire
Name Note 0.5mm (0.02 in.) dia., Positive leg Type R
MDL Number MFCD00198186
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13018BY

  SDS
alfa aesar™
13018BY
100cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note 0.5mm (0.02 in.) dia., Positive leg Type R
Odor Odorless
Weight 19.595g/cc
Chemical Name or Material Platinum 13 wt% Rhodium wire
Physical Form Wire
TSCA TSCA
MDL Number MFCD00198186
Quantity 100cm
Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 87:13 wt%