missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum 13 wt% Rhodium wire, 0.25mm (0.01 in.) dia., ISA Type R Standard Grade Thermocouple

For ISA Type R Standard Grade Thermocouple

Specifications

Chemical Name or Material Platinum 13 wt% Rhodium wire
Name Note 0.25mm (0.01 in.) dia.
MDL Number MFCD00198186
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10062BU View Documents Alfa Aesar™
10062BU
25cm N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA10062BY View Documents Alfa Aesar™
10062BY
100cm N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA10062CC View Documents Alfa Aesar™
10062CC
500cm N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

Platinum 13 wt% Rhodium wire
0.25mm (0.01 in.) dia.
MFCD00198186