Alfa Aesar™ Platinum 13 wt% Rhodium wire, 0.25mm (0.01 in.) dia., ISA Type R Standard Grade Thermocouple
For ISA Type R Standard Grade Thermocouple
Manufacturer:  Alfa Aesar™ 10062CC
 View more versions of this product
Catalog No. AA10062CC
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Pt:Rh; 87:13 wt%
≈0.26g/25cm
0.25mm (0.01 in.) dia.
Odorless
Platinum 13 wt% Rhodium wire
500cm
MFCD00198186
Wire, ≈0.26g/25cm
Documents