Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum 13 wt% Rhodium wire, 0.203mm (0.008 in.) dia., ISA Type R Standard Grade Thermocouple

$332.49 - $332.49

Chemical Identifiers

Chemical Name or Material Platinum 13 wt% Rhodium wire
Name Note 0.203mm (0.008 in.) dia., ISA Type R Standard Grade Thermocouple
MDL Number MFCD00198186
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42905G1

 
alfa aesar™
42905G1
1m Each for $332.49
Description & Specifications

Specifications

Chemical Name or Material Platinum 13 wt% Rhodium wire
TSCA TSCA
Name Note 0.203mm (0.008 in.) dia., ISA Type R Standard Grade Thermocouple
Physical Form Wire
MDL Number MFCD00198186
Odor Odorless
Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 87:13 wt%
Weight ≈0.64g/m
Quantity 1m