Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum 10 wt% Rhodium wire, 0.5mm (0.02 in.) dia., ISA Type S Standard Grade Thermocouple

For ISA Type S Standard Grade Thermocouple

Chemical Identifiers

Chemical Name or Material Platinum 10 wt% Rhodium wire
Name Note 0.5mm (0.02 in.) dia.
MDL Number MFCD00198186
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13372BY

  SDS
alfa aesar™
13372BY
100cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum 10 wt% Rhodium wire
TSCA TSCA
Name Note 0.5mm (0.02 in.) dia.
Physical Form Wire
MDL Number MFCD00198186
Odor Odorless
Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 90:10 wt%
Weight ≈1.04g/25cm
Quantity 100cm