missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum 10 wt% Rhodium wire, 0.25mm (0.01 in.) dia., ISA Type S Standard Grade Thermocouple

For ISA Type S Standard Grade Thermocouple

Specifications

Chemical Name or Material Platinum 10 wt% Rhodium wire
Name Note 0.25mm (0.01 in.) dia.
MDL Number MFCD00198186
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10065BY
View Documents
Alfa Aesar™
10065BY
100cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA10065BU
View Documents
Alfa Aesar™
10065BU
25cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA10065CC
View Documents
Alfa Aesar™
10065CC
500cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Specifications

Platinum 10 wt% Rhodium wire
0.25mm (0.01 in.) dia.
MFCD00198186
SDS