missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum 10 wt% Rhodium wire, 0.25mm (0.01 in.) dia., ISA Type S Standard Grade Thermocouple

For ISA Type S Standard Grade Thermocouple

Manufacturer:  Alfa Aesar™ 10065BY

 View more versions of this product

Catalog No. AA10065BY

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Pt:Rh; 90:10 wt%
≈0.52g/50cm
0.25mm (0.01 in.) dia.
Insoluble in water.
Wire, ≈0.52g/50cm
Platinum 10 wt% Rhodium wire
100cm
MFCD00198186
Odorless
SDS
Documents