Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 10% on activated carbon powder, standard, reduced, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Platinum
Name Note 10% on activated carbon powder, Standard, reduced, nominally 50% water wet
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3832506

  SDS
alfa aesar™
3832506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3832514

  SDS
alfa aesar™
3832514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3832522

  SDS
alfa aesar™
3832522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum
Name Note 10% on activated carbon powder, Standard, reduced, nominally 50% water wet
TSCA TSCA
MDL Number MFCD00011179
Odor Odorless
Quantity 5g
Weight 900 to 1100m2/g

  • Aromatic ring hydrogenation, aliphatic aldehydes and ketones to alcohols, conversion of -CH=CH- to -CH2-CH2-