Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 10% on activated carbon paste, Type

Chemical Identifiers

Chemical Name or Material Platinum
CAS 7440-06-4
Name Note 10% on activated carbon paste, Type 128M
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3604406

  SDS
alfa aesar™
H3604406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3604414

  SDS
alfa aesar™
H3604414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3604422

  SDS
alfa aesar™
H3604422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum
TSCA TSCA
Odor Odorless
Name Note 10% on activated carbon paste, Type 128M
Physical Form Solid
MDL Number MFCD00011179
CAS 7440-06-4
Color Black
Quantity 5g

Applications
Platinum as a catalyst is used for hydrogenation of alkenes, alkynes, nitro groups. It is also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

Solubility
Insoluble in water, mineral acids, and organic acids. Soluble in boiling aqua regia (reacts), and molten alkali cyanides (reacts).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, organic materials and flammable substances.