Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 1% on activated carbon powder, standard, unreduced, nominally 50% water wet

Schiff's base and nitrohydrogenation, hydrosilation

Chemical Identifiers

Chemical Name or Material Platinum
Name Note 1% on activated carbon powder, Standard, unreduced, nominally 50% water wet
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3831306

  SDS
alfa aesar™
3831306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3831314

  SDS
alfa aesar™
3831314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3831322

  SDS
alfa aesar™
3831322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Platinum
Weight 900 to 1100m2/g
TSCA TSCA
MDL Number MFCD00011179
Name Note 1% on activated carbon powder, Standard, unreduced, nominally 50% water wet