Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 1% on activated carbon powder, standard, reduced, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Platinum
Name Note 1% on activated carbon powder, Standard, reduced, nominally 50% water wet
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3831214

 
alfa aesar™
3831214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3831222

 
alfa aesar™
3831222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum
TSCA TSCA
Name Note 1% on activated carbon powder, Standard, reduced, nominally 50% water wet
MDL Number MFCD00011179
Quantity 25g
Weight 900 to 1100m2/g