Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 1% on activated carbon paste, Type 163

Chemical Identifiers

Chemical Name or Material Platinum
CAS 7440-06-4
Name Note 1% on activated carbon paste, Type 163
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3683906

  SDS
alfa aesar™
H3683906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3683914

  SDS
alfa aesar™
H3683914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3683922

  SDS
alfa aesar™
H3683922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum
TSCA TSCA
Odor Odorless
Name Note 1% on activated carbon paste, Type 163
Physical Form Solid
MDL Number MFCD00011179
CAS 7440-06-4
Color Dark gray
Quantity 5g