missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Platinum, 1% on activated carbon paste, 1R18MA, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 4551914
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Platinum | |
7440-06-4 | |
25 g | |
MFCD00011179 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 |
1% on activated carbon paste, 1R18MA | |
Paste | |
Pt | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
[Pt] | |
195.08 | |
CHEBI:33400 |
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
EINECSNumber : 231-116-1
TSCA : No
Recommended Storage : Ambient temperatures