missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 0.5% on 2.7-3.3mm (0.11-0.13in) alumina pellets, reduced

Manufacturer:  Alfa Aesar™ 8910636

 View more versions of this product

Catalog No. AA8910636



Description

Description

  • Selective hydrogenations; gas purifications: e
g
  • Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide
  • Specifications

    Specifications

    Platinum
    0.5% on alumina pellets, 2.7 to 3.3mm (0.11 to 0.13 in.), Reduced
    MFCD00011179
    500g
    7440-6-6

    Chemical Identifiers

    7440-6-6
    MFCD00011179
    Safety and Handling

    Safety and Handling

    TSCA : TSCA

    SDS Documents