Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 0.3% on 2.7-3.3mm (0.11-0.13in) alumina pellets, reduced

Chemical Identifiers

Chemical Name or Material Platinum
Name Note 0.3% on 2.7 to 3.3mm (0.11 to 0.13 in.) alumina pellets, Reduced
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3982614

  SDS
alfa aesar™
3982614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3982622

  SDS
alfa aesar™
3982622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum
TSCA TSCA
Name Note 0.3% on 2.7 to 3.3mm (0.11 to 0.13 in.) alumina pellets, Reduced
MDL Number MFCD00011179
Quantity 25g

  • Selective hydrogenation, gas purifications: e.g. Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide