Learn More
Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04150814
Description
Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst.Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane is used as a catalyst in hydrosilylation reaction for the preparation of trans-beta-silanes and in reductive amination reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst
Specifications
Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex | |
68478-92-2 | |
0.984 g/mL | |
C8 H18 O Pt Si2 | |
25 g | |
platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, platinum salt, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution, ksc628a5r, platinum 1,3-divinyl-1,1,3,3-tetramethyldisiloxane, dimethyl vinyl silyl oxy-dimethyl-vinyl-silane; platinum, ethenyl ethenyldimethylsilyl oxy dimethylsilane platinum, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, platinum,1,3-diethenyl-1,1,3,3-tetramethyldisiloxane complexes | |
RCNRJBWHLARWRP-UHFFFAOYSA-N | |
ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane;platinum | |
10959889 | |
Liquid |
Soln. in vinyl terminated polydimethylsiloxane | |
Orange | |
>110°C (230°F) | |
MFCD00151662 | |
Moisture sensitive | |
Miscible with toluene,xylene,isopropyl alcohol,double-vinyl silicone oil and vinyl terminated poly(dimethylsiloxane). Immiscible with water. | |
[Pt].C[Si](C)(O[Si](C)(C)C=C)C=C | |
474.68 | |
474.68 |
Chemical Identifiers
68478-92-2 | |
474.68 | |
RCNRJBWHLARWRP-UHFFFAOYSA-N | |
10959889 |
C8 H18 O Pt Si2 | |
MFCD00151662 | |
platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, platinum salt, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution, ksc628a5r, platinum 1,3-divinyl-1,1,3,3-tetramethyldisiloxane, dimethyl vinyl silyl oxy-dimethyl-vinyl-silane; platinum, ethenyl ethenyldimethylsilyl oxy dimethylsilane platinum, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, platinum,1,3-diethenyl-1,1,3,3-tetramethyldisiloxane complexes | |
[Pt].C[Si](C)(O[Si](C)(C)C=C)C=C |
Safety and Handling
H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only