Please login to your online account to display your discounted pricing

Platinizing Solution, For Electrical Conductivity and Resistivity, LabChem™

$1,424.83 - $1,424.83

Chemical Identifiers

Chemical Name or Material Platinizing Solution
CAS 7732-18-5
CAS Max % 96.7
CAS 18497-13-7
CAS Max % 3
CAS 6080-56-4
CAS Max % 0.3
Name Note For Electrical Conductivity and Resistivity
Linear Formula PtCl4·2HCl·6H2O
Formula Weight 517.9
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC186807

 
lab chem
LC186807
125mL Each for $1,424.83
Description & Specifications

Specifications

Formula Weight 517.9
CAS 6080-56-4
Specific Gravity 1g/mL
Solubility Soluble in water
Quantity 125mL
CAS Max % 96.7
Packaging Amber Glass
Storage Information Room Temperature
Identification Passes Test
Decomposition Information Hydrogen chloride; Lead oxide
Health Hazard 2 GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage fertility or the unborn child.
Harmful to aquatic life with long lasting effects.
Linear Formula PtCl4·2HCl·6H2O
Chemical Name or Material Platinizing Solution
Physical Form Liquid
Health Hazard 1 Danger
CAS 18497-13-7
CAS Max % 3
CAS 7732-18-5
Name Note For Electrical Conductivity and Resistivity
CAS Max % 0.3
Density 1g/mL
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist.
Wear protective gloves, eye protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Color Clear, yellow orange
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist.
Wear protective gloves, eye protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage fertility or the unborn child.
Harmful to aquatic life with long lasting effects.