Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Plastin-L Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Plastin-L Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1326P1ABX

 View more versions of this product

Catalog No. MS1326P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Format Concentrated
Isotype IgG1
Quantity 100µL
Immunogen Human recombinant L-plastin protein
Regulatory Status RUO
Clone LPL4A.1
Cross Reactivity Human
Antigen Plastin-L Ab-1
Research Discipline Cytoskeletal Protein
Type Primary
Concentration 1mg/mL
Conjugate Unlabeled
Classification Monoclonal

Provide accurate, reproducible results with the Thermo Scientific™ Plastin-L Ab-1, Mouse Monoclonal Antibody.

L-plastin is a leukocyte specific actin bundling protein from fimbrin family of proteins. It has two actin-binding domains and two EF hand type calcium-binding domains. It gets phosphorylated on the serine residue in its headpiece region in response to a variety of inflammatory stimuli. It is hypothesized that L-plastin may play a role in regulating integrin mediated adhesion in leukocytes. L-plastin is expressed in majority of human cancer cell lines derived from solid tumors. Its expression is positively upregulated by testosteronse in andrgen receptor positive prostrate and breast cancer cells through three androgen receptor binding elements located in the upstream region of the gene.

Host Species: Mouse

Clone: LPL4A.1

Isotype: IgG1

Species Reactivity: Human. Does not react with Mouse. Others not known.

Epitope: Headpiece region

Immunogen: Human recombinant L-plastin protein

Molecular Weight: 62kDa

Positive Control: Human polymorphonuclear neutrophils and monocytes, MCF-7, Jurkat cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence (Fixed in methanol or 3% paraformaldehyde)
  • Immunoprecipitation (Native)

Note:

USA: RUO; Int'l: RUO