Please login to your online account to display your discounted pricing

Thermo Scientific™ Plastic Spill-Free Slide Jar

Hold 10 slides, one per slot with the Thermo Scientific™ Plastic Spill-Free Slide Jar.

$574.33 - $574.33

Specifications

Length (English) 1.4 in.
Length (Metric) 36mm
Slide Count 10
Width (English) 1 in.
Width (Metric) 26mm
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

10-013-63

 
thermo scientific™
1001363
12/Pack Case of 12 Each for $574.33
Description & Specifications

Specifications

Length (English) 1.4 in.
Length (Metric) 36mm
Slide Count 10
Width (English) 1 in.
Width (Metric) 26mm

Hold 10 slides, one per slot with the Thermo Scientific™ Plastic Spill-Free Slide Jar.

  • 1.4 x 1″ (36 x 26mm) rectangular top with straight body
  • Polypropylene construction with internal grooves
  • Integral snap-lock lid with reagent-proof lid seal
  • Microwavable