Learn More
Eisco™ Plastic Filter Funnels
Eisco™ Plastic Filter Funnels are constructed of polyethylene which is chemical, acid, and alkaline resistant.
Supplier:  Eisco™ CH0454D10
 View more versions of this product
Catalog No. S23821
Only null left
Explore available promotions
Description
Description
  • Chemically resistant polyethylene funnel
  • Translucent
  • Durable for student use
  • Easy to clean
  • Autoclavable
Warranty
1 year limited manufacturer warranty.
Specifications
Specifications
100 mm
Polyethylene
Clear
6 in.
10 cm
15.2 cm
10 Pk.
Filtering Funnel
Product Suggestions
Product Suggestions