American Educational Products Plastic Column Kit
Manufacturer:  American Educational Products HUB3180
 View more versions of this product
Catalog No. S53016A
Includes: tube, drainage fixture, screen, and teacher's guide.
Description
Description
Learn how the movement of water is affected by substrate!
How do porosity and permeability affect the distribution of aquifers? Help students study these important concepts and integrate math measurement and percentage skills. Bead set is optional. Grades 6-12.
Specifications
Specifications
Plastic Column Kit
12
6
Education
Documents