American Educational Products Plastic Column Kit
Manufacturer:  American Educational Products HUB3275
 View more versions of this product
Catalog No. S53036
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes: tube, drainage fixture, screen, and teacher's guide.
Description
Description
Learn how the movement of water is affected by substrate!
How do porosity and permeability affect the distribution of aquifers? Help students study these important concepts and integrate math measurement and percentage skills. Bead set is optional. Grades 6-12.
Specifications
Specifications
12
6
4mm Beads