Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Plasminogen Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Plasminogen Ab-1, Mouse Monoclonal Antibody. 

$268.92 - $1,588.41

Specifications

Antigen Plasminogen Ab-1
Clone SBF1
Applications Western Blot
Classification Monoclonal
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS659B

 
thermo scientific™ lab vision™
MS659B
200μg/mL Biotin 1mL Each for $1,588.41

MS659B0

 
thermo scientific™ lab vision™
MS659B0
200μg/mL Biotin 100μL Each for $364.29

MS659B1

 
thermo scientific™ lab vision™
MS659B1
200μg/mL Biotin 500μL Each for $991.04

MS659P

 
thermo scientific™ lab vision™
MS659P
200μg/mL Unlabeled 1mL Each for $1,259.18

MS659P0

 
thermo scientific™ lab vision™
MS659P0
200μg/mL Unlabeled 100μL Each for $268.92

MS659P1

 
thermo scientific™ lab vision™
MS659P1
200μg/mL Unlabeled 500μL Each for $761.65

MS659P1ABX

 
thermo scientific™ lab vision™
MS659P1ABX
1mg/mL Unlabeled 100μL Each for $846.39

MS659PABX

 
thermo scientific™ lab vision™
MS659PABX
1mg/mL Unlabeled 200μL Each for $1,383.49
Description & Specifications

Specifications

Antigen Plasminogen Ab-1
Clone SBF1
Applications Western Blot
Classification Monoclonal
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Hematopoietic
Type Primary
Immunogen Human plasminogen protein
Isotype IgG2a kappa

Provide accurate, reproducible results with the Thermo Scientific™ Plasminogen Ab-1, Mouse Monoclonal Antibody.

Plasminogen is a key proenzyme of plasmin in the fibrinolytic and thrombolytic systems, the deficiency of which leads to a mild thrombotic tendency. Plasminogen binds to fibrin as well as to the surface of endothelial cells and monocytes/macrophages, where it is activated to plasmin. Streptokinase is a plasminogen activator widely used in treating blood-clotting disorders. Complexes of streptokinase with human plasminogen can hydrolytically activate other plasminogen molecules to plasmin, which then dissolves blood clots. Apolipoprotein(a) is very similar to plasminogen in terms of amino acid sequence, it inhibits plasmin generation by competing with plasminogen's binding to fibrin and endothelial cells.

Host Species: Mouse

Clone: SBF1

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human plasminogen protein

Molecular Weight: ∽120kDa

Positive Control: HUVEC cells

Cellular Localization: Secreted protein

Recommended for:

  • Western Blotting (Nonreduced only)

Note:

USA: RUO; Int'l: RUO