Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Plasminogen Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Plasminogen Ab-1, Mouse Monoclonal Antibody. 

Manufacturer: thermo scientific™ lab vision™  MS659P1ABX

 View more versions of this product

Catalog No. MS659P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Isotype IgG2a kappa
Clone SBF1
Host Species Mouse
Immunogen Human plasminogen protein
Quantity 100µL
Antigen Plasminogen Ab-1
Applications Western Blotting
Format Concentrated
Regulatory Status RUO
Classification Monoclonal
Conjugate Unlabeled
Research Discipline Hematopoietic
Concentration 1mg/mL
Type Primary

Provide accurate, reproducible results with the Thermo Scientific™ Plasminogen Ab-1, Mouse Monoclonal Antibody.

Plasminogen is a key proenzyme of plasmin in the fibrinolytic and thrombolytic systems, the deficiency of which leads to a mild thrombotic tendency. Plasminogen binds to fibrin as well as to the surface of endothelial cells and monocytes/macrophages, where it is activated to plasmin. Streptokinase is a plasminogen activator widely used in treating blood-clotting disorders. Complexes of streptokinase with human plasminogen can hydrolytically activate other plasminogen molecules to plasmin, which then dissolves blood clots. Apolipoprotein(a) is very similar to plasminogen in terms of amino acid sequence, it inhibits plasmin generation by competing with plasminogen's binding to fibrin and endothelial cells.

Host Species: Mouse

Clone: SBF1

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human plasminogen protein

Molecular Weight: ∽120kDa

Positive Control: HUVEC cells

Cellular Localization: Secreted protein

Recommended for:

  • Western Blotting (Nonreduced only)

Note:

USA: RUO; Int'l: RUO