Alfa Aesar™ Plasmid DNA isolation solution III
Specifications
Chemical Name or Material Plasmid DNA isolation solution III
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60196AP View Documents Alfa Aesar™
J60196AP
500mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ60196AK View Documents Alfa Aesar™
J60196AK
250mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
This solution returns the pH to neutral after lysis with SDS and NaOH and allows DNA strands to renature and partially rehybridize. At the same time, the potassium acetate precipitates the detergent from the cell suspension.
Specifications
Specifications
Plasmid DNA isolation solution III