Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Plasma Cell Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Plasma Cell Ab-1, Mouse Monoclonal Antibody.

$238.04 - $947.06

Specifications

Antigen Plasma Cell Ab-1
Clone LIV3G11
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS488P

 
thermo scientific™ lab vision™
MS488P
Purified Ab with BSA and Azide 1mL Each for $828.78

MS488P0

 
thermo scientific™ lab vision™
MS488P0
Purified Ab with BSA and Azide 100μL Each for $238.04

MS488P1

 
thermo scientific™ lab vision™
MS488P1
Purified Ab with BSA and Azide 500μL Each for $649.14

MS488P1ABX

 
thermo scientific™ lab vision™
MS488P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $729.22

MS488PABX

 
thermo scientific™ lab vision™
MS488PABX
Ab without BSA/Azide (Frozen) 200μL Each for $947.06

MS488R7

 
thermo scientific™ lab vision™
MS488R7
Ready-to-use for IHC (UVLP) 7mL Each for $567.94
Description & Specifications

Specifications

Antigen Plasma Cell Ab-1
Clone LIV3G11
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Murine
Regulatory Status RUO
Research Discipline Hematopoietic
Type Primary
Isotype IgG2a

Provide accurate, reproducible results with the Thermo Scientific™ Plasma Cell Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: LIV3G11

Isotype: IgG2a

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Positive Control: Tonsil or lymph node

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO