Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Plasma Cell Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Plasma Cell Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS488P1ABX

 View more versions of this product

Catalog No. MS488P1ABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Quantity 100µL
Isotype IgG2a
Clone LIV3G11
Regulatory Status RUO
Antigen Plasma Cell Ab-1
Host Species Mouse
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Conjugate Unlabeled
Type Primary
Research Discipline Hematopoietic
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Classification Monoclonal

Provide accurate, reproducible results with the Thermo Scientific™ Plasma Cell Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: LIV3G11

Isotype: IgG2a

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Positive Control: Tonsil or lymph node

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO