missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Plas Labs™ Model 830-CP Compact Glove Box

Plas Lab Model 830-CP Compact Glove Box is designed specifically for use when weighing hazardous materials that must be contained.

Manufacturer:  Plas Labs™ 830CP/20

 View more versions of this product

Catalog No. 11-386-140

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1 Year
29 in.
73.66cm
Glove Box
49 in.
Determining the dry weight or moisture content of aqueous solutions, adhesives, cereals, toxic chemicals, pharmaceuticals, paper, plastics and light radioactive materials
79cm
35 in.
91.44cm
UL, CSA, CE
78.74cm
30 in.
31 in.
HEPA, Organic Vapor
30 in.
17.6 cu. ft.
125cm