Please login to your online account to display your discounted pricing

Plas Labs™ Model 830-CP Compact Glove Box


Plas Lab Model 830-CP Compact Glove Box is designed specifically for use when weighing hazardous materials that must be contained.

Manufacturer: plas labs™  830CP/11

 View more versions of this product

Catalog No. 11-386-139

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Warranty 1 Year
Certifications/Compliance UL, CSA, CE
Depth (English) Interior 23 in.
Depth (Metric) Exterior 76.2cm
Depth (Metric) Interior 58.42cm
Depth (English) Exterior 31 in.
Type Glove Box
Height (English) Exterior 34 in.
Width (English) Exterior 63 in.
Filter Type HEPA, Organic Vapor
For Use With (Application) Determining the dry weight or moisture content of aqueous solutions, adhesives, cereals, toxic chemicals, pharmaceuticals, paper, plastics and light radioactive materials
Height (English) Interior 30 in.
Height (Metric) Exterior 86cm
Volume (English) 11.2 cu. ft.
Width (English) Interior 29 in.
Width (Metric) Exterior 160cm
Width (Metric) Interior 73.6cm