Please login to your online account to display your discounted pricing

Plas Labs™ Model 830-CP Compact Glove Box


Plas Lab Model 830-CP Compact Glove Box is designed specifically for use when weighing hazardous materials that must be contained.

Manufacturer: plas labs™  830CP/30

 View more versions of this product

Catalog No. 11-386-142

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Warranty 1 Year
Width (Metric) Interior 101.6cm
Height (Metric) Exterior 79cm
Type Glove Box
Height (English) Interior 32 in.
Width (English) Interior 40 in.
Width (English) Exterior 49 in.
Width (Metric) Exterior 125cm
Certifications/Compliance UL, CSA, CE
Depth (Metric) Exterior 78.74cm
Depth (English) Exterior 30 in.
Volume (English) 25.8 cu. ft.
Filter Type HEPA, Organic Vapor
For Use With (Application) Determining the dry weight or moisture content of aqueous solutions, adhesives, cereals, toxic chemicals, pharmaceuticals, paper, plastics and light radioactive materials
Depth (Metric) Interior 73.66cm
Height (English) Exterior 31 in.
Depth (English) Interior 29 in.