missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Plant Trays

Perfect seed starters that never rot or rust.

Manufacturer:  Fisher Scientific GRS812

 View more versions of this product

Catalog No. S17642




Description

Description

 

  • Made of high-impact grey polystyrene
  • Can also be used for a variety of laboratory applications
  • Nest for easy storage

 

Specifications

Specifications

Grey
6.35cm
Used for a variety of laboratory applications
20.3cm
Plant Tray
12 in.
2.5 in.
8 x 12 in. (20.3 x 30.4cm)
8 in.
Polystyrene
Each
30.4cm
Documents