Please login to your online account to display your discounted pricing

Placental Alkaline Phosphatase (PLAP) Mouse anti-Human, Biotin, Clone: 8B6, eBioscience™

Mouse Monoclonal Antibody

$140.65 - $353.80

Specifications

Antigen Placental Alkaline Phosphatase (PLAP)
Clone 8B6
Host Species Murine
Gene Alias PLAP
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122619

 
life technologies
LS13987080
25μg Each for $140.65

501122620

 
life technologies
LS13987082
100μg Each for $353.80
Description & Specifications

Specifications

Antigen Placental Alkaline Phosphatase (PLAP)
Clone 8B6
Host Species Murine
Gene Alias PLAP
Species Reactivity Human
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 8B6 recognizes human Placental Alkaline Phosphatase (PLAP), a 130 (or 130 with 65 kDa) enzyme expressed by placental syncytiotrophoblastoid cells. This enzyme has slightly different enzyme kinetics from other phosphatases. PLAP can be found anchored to the plasma membrane in addition to the cytoplasm. Expression is found in several germ line cancers; ovarian and testicular. Expression is also present in human embryonic stem cells and bone marrow-derived mesenchymal stromal cells.

This 8B6 antibody reacts with both the Regan and Nagao isozymes.