missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Placental Alk. Phosphatase (PLAP) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Placental Alk. Phosphatase (PLAP) Ab-1, Mouse Monoclonal Antibody.

$336.86 - $1,172.14

Specifications

Antigen Placental Alk. Phosphatase (PLAP) Ab-1
Clone 8B6.18
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Murine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
MS208B Thermo Scientific™ Lab Vision™
MS208B
200μg/mL Biotin 1mL Each for $1,172.14
MS208B0 Thermo Scientific™ Lab Vision™
MS208B0
200μg/mL Biotin 100μL Each for $336.86
MS208B1 Thermo Scientific™ Lab Vision™
MS208B1
200μg/mL Biotin 500μL Each for $880.53
MS208P Thermo Scientific™ Lab Vision™
MS208P
200μg/mL Unlabeled 1mL Each for $835.00
MS208P1 Thermo Scientific™ Lab Vision™
MS208P1
200μg/mL Unlabeled 500μL Each for $661.33
MS208P1ABX Thermo Scientific™ Lab Vision™
MS208P1ABX
1mg/mL Unlabeled 100μL Each for $752.04
MS208PABX Thermo Scientific™ Lab Vision™
MS208PABX
1mg/mL Unlabeled 200μL Each for $944.86
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Placental Alk. Phosphatase (PLAP) Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 8B6.18

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP)

Molecular Weight: 60-70kDa

Positive Control: Placenta

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • ELISA

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Placental Alk. Phosphatase (PLAP) Ab-1
Immunohistochemistry (Paraffin)
Murine
Cancer and Tumor Biology
HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP)
8B6.18
Monoclonal
RUO
Primary
IgG2a kappa