Please login to your online account to display your discounted pricing

PK 11195, Tocris

Antagonist at peripheral benzodiazepine receptors

$113.57 - $478.51

Chemical Identifiers

Chemical Name or Material PK 11195
Assay Percent Range >98%
Molecular Formula C21H21ClN2O
Formula Weight 352.86
Synonym 1-(2-Chlorophenyl)-N-methyl-N-(1-methylpropyl)-3-isoquinolinecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

067010

 
r & d systems
RDS067010
10mg Each for $113.57

067050

 
r & d systems
RDS067050
50mg Each for $478.51
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 352.86
Assay Percent Range >98%
Synonym 1-(2-Chlorophenyl)-N-methyl-N-(1-methylpropyl)-3-isoquinolinecarboxamide
Recommended Storage Store at Room Temperature
Chemical Name or Material PK 11195
Molecular Formula C21H21ClN2O
CAS 85532-75-8
Safety and Handling

Store at Room Temperature