Thermo Scientific™ pJET1.2 Sequencing Primers
Primers for sequencing of DNA fragments inserted into Eco32I site within the eco47IR gene of pJET1.2 and for colony screening by PCR.
$48.47 - $48.85
Specifications
Quantity 0.2 A260, (6.7µg)
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
FERSO511 View Documents Thermo Scientific™
SO511
pJET™1.2 forward sequencing primer, 24-mer Each for $48.47
FERSO501 View Documents Thermo Scientific™
SO501
pJET™1.2 forward sequencing primer, 23-mer Each for $48.85
Description
Description
Thermo Scientific sequencing primers are single-stranded oligonucleotides with 5'-hydroxyl and 3'-hydroxyl ends. pJET1.2 sequencing primers flank the Eco32I site in the eco47IR gene of positive selection cloning vector pJET1.2. All primers are supplied as 10 µM aqueous solutions.
Applications
  • Sequencing of DNA fragments inserted into Eco32I site within the eco47IR gene of pJET1.2
  • Colony screening by PCR
Specifications
Specifications
0.2 A260, (6.7µg)