missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pizotifen, TCI America™

Manufacturer:  TCI America P234450MG

 View more versions of this product

Catalog No. P234450MG



Specifications

Specifications

Pizotifen
149°C
15574-96-6
MFCD00864192
FIADGNVRKBPQEU-UHFFFAOYSA-N
4-(4,5-dihydrobenzo[1,2]cyclohepta[3,4-b]thiophen-10-ylidene)-1-methylpiperidine
27400
295.44
≥98.0% (HPLC)
White
50mg
C19H21NS
4-[9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene]-1-methylpiperidine, Sandomigran
CN1CCC(=C2C3=C(CCC4=CC=CC=C42)SC=C3)CC1
295.444
CHEBI:50212
Crystal-Powder at 20°C

Chemical Identifiers

15574-96-6
295.444
FIADGNVRKBPQEU-UHFFFAOYSA-N
27400
4-(4,5-dihydrobenzo[1,2]cyclohepta[3,4-b]thiophen-10-ylidene)-1-methylpiperidine
C19H21NS
MFCD00864192
4-[9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene]-1-methylpiperidine, Sandomigran
CHEBI:50212
CN1CCC(=C2C3=C(CCC4=CC=CC=C42)SC=C3)CC1
Safety and Handling

Safety and Handling

RTECSNumber : TM7165000

TSCA : No

Documents