missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pivaloylacetonitrile, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 418020250

 View more versions of this product

Catalog No. AC418020250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Pivaloylacetonitrile
Glass bottle
White to Cream
25g
96% min. (GC)
(CH3)3CC(O)CH2CN
1-cyano-3,3-dimethyl-2-butanone, 4,4-dimethyl-3-oxo-pentanenitrile, 4,4-dimethyl-3-oxovaleronitrile, cyanopinacolone, pentanenitrile, 4,4-dimethyl-3-oxo, pivaloyl acetonitrile, pivaloylacetonitrile, pyvaolylacetonitrile, trimethylacetylacetonitrile, valeronitrile, 4,4-dimethyl-3-oxo
CC(C)(C)C(=O)CC#N
125.17
125.17
97%
Authentic
1% max. (K.F.)
66°C to 72°C
59997-51-2
C7H11NO
MFCD00010208
MXZMACXOMZKYHJ-UHFFFAOYSA-N
4,4-dimethyl-3-oxopentanenitrile
108871
Crystals or Crystalline Powder

Chemical Identifiers

59997-51-2
125.17
MXZMACXOMZKYHJ-UHFFFAOYSA-N
108871
CC(C)(C)C(=O)CC#N
C7H11NO
MFCD00010208
1-cyano-3,3-dimethyl-2-butanone, 4,4-dimethyl-3-oxo-pentanenitrile, 4,4-dimethyl-3-oxovaleronitrile, cyanopinacolone, pentanenitrile, 4,4-dimethyl-3-oxo, pivaloyl acetonitrile, pivaloylacetonitrile, pyvaolylacetonitrile, trimethylacetylacetonitrile, valeronitrile, 4,4-dimethyl-3-oxo
4,4-dimethyl-3-oxopentanenitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 262-017-1

SDS Documents