Please login to your online account to display your discounted pricing

Pivaldehyde, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC368360050

 View more versions of this product

Catalog No. AC368360050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 86.13
Quantity 5g
Color Clear colorless
Beilstein 01, IV, 3295
Fieser 13,249
Merck Index 15, 7624
Density 0.7750g/mL
Infrared Spectrum Authentic
Chemical Name or Material Pivaldehyde
Assay Percent Range 97%
Molecular Formula C5H10O
Linear Formula (CH3)3CCHO
MDL Number MFCD00006962
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Name Note 98%
CAS 630-19-3
Synonym Trimethylacetaldehyde
Refractive Index 1.3775 to 1.3795 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Pivaldehyde
Name Note 98%
CAS 630-19-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H10O
Linear Formula (CH3)3CCHO
Formula Weight 86.13
MDL Number MFCD00006962
Synonym Trimethylacetaldehyde
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents