missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pivaldehyde, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC368361000

 View more versions of this product

Catalog No. AC368361000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Pivaldehyde
−15°C
Glass bottle
0.775
Colorless
100g
630-19-3
C5H10O
MFCD00006962
13,249
2,2-dimethylpropionaldehyde, neopentanal, pivalaldehyde, pivaldehyde, pivalic aldehyde, propanal, 2,2-dimethyl, trimethyl acetaldehyde, trimethylacetaldehyde, trimethylacetaldehyde pivaldehyde, unii-ssc20260qw
FJJYHTVHBVXEEQ-UHFFFAOYSA-N
2,2-dimethylpropanal
12417
Liquid
0.7750g/mL
Authentic
1.3775 to 1.3795
75.0°C
6.0°C
98%
95% min. (GC)
(CH3)3CCHO
01, IV, 3295
15, 7624
Solubility in water: negligibly soluble.
CC(C)(C)C=O
86.13
86.13
96%

Chemical Identifiers

630-19-3
86.13
FJJYHTVHBVXEEQ-UHFFFAOYSA-N
12417
CC(C)(C)C=O
C5H10O
MFCD00006962
2,2-dimethylpropionaldehyde, neopentanal, pivalaldehyde, pivaldehyde, pivalic aldehyde, propanal, 2,2-dimethyl, trimethyl acetaldehyde, trimethylacetaldehyde, trimethylacetaldehyde pivaldehyde, unii-ssc20260qw
2,2-dimethylpropanal
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-134-6

SDS Documents