missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pivalamidine Hydrochloride, TCI America™

Manufacturer:  TCI America P176125G

 View more versions of this product

Catalog No. P176125G



Specifications

Specifications

Pivalamidine Hydrochloride
≥98.0% (N,T)
MFCD00051988
ARDGQYVTLGUJII-UHFFFAOYSA-N
2,2-dimethylpropanimidamide;hydrochloride
2781880
Crystal-Powder at 20°C
193°C
18202-73-8
C5H13ClN2
2,2-Dimethylpropionamidine Hydrochloride
CC(C)(C)C(=N)N.Cl
136.623
136.62
White-Yellow
25g

Chemical Identifiers

18202-73-8
136.623
ARDGQYVTLGUJII-UHFFFAOYSA-N
2781880
CC(C)(C)C(=N)N.Cl
C5H13ClN2
MFCD00051988
2,2-Dimethylpropionamidine Hydrochloride
2,2-dimethylpropanimidamide;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents