Please login to your online account to display your discounted pricing

PIT, Tocris

P2Y ligand; displays mixed antagonism/potentiation

$146.74 - $618.87

Chemical Identifiers

Chemical Name or Material PIT
Assay Percent Range >99%
Molecular Formula C13H8N2O2·C7H8O3S
Formula Weight 396.42
Synonym 2-(2-Pyridinyl)-(3H)-indol-3-one-1-oxide 4-methylbenzenesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

168250

 
r & d systems
RDS168250
50mg Each for $618.87

168210

 
r & d systems
RDS168210
10mg Each for $146.74
Description & Specifications

Specifications

Formula Weight 396.42
Quantity 50mg
CAS 56583-49-4
Assay Percent Range >99%
Synonym 2-(2-Pyridinyl)-(3H)-indol-3-one-1-oxide 4-methylbenzenesulfonate
Molecular Formula C13H8N2O2·C7H8O3S
Chemical Name or Material PIT
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C