Please login to your online account to display your discounted pricing

PIT 1, Tocris™

Inhibits Akt signaling

$199.70 - $834.54

Chemical Identifiers

Chemical Name or Material PIT 1
Assay Percent Range >99%
Molecular Formula C14H10ClN3O4S
Formula Weight 351.76
Synonym N-[[(3-Chloro-2-hydroxy-5-nitrophenyl)amino]thioxomethyl]benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

416850

 
r & d systems
RDS416850
50mg Each for $834.54

416810

 
r & d systems
RDS416810
10mg Each for $199.70
Description & Specifications

Specifications

Formula Weight 351.76
Quantity 50mg
Chemical Name or Material PIT 1
Assay Percent Range >99%
Molecular Formula C14H10ClN3O4S
CAS 53501-41-0
Synonym N-[[(3-Chloro-2-hydroxy-5-nitrophenyl)amino]thioxomethyl]benzamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C