missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Piroxicam, TCI America™

Manufacturer:  TCI America P19052G

 View more versions of this product

Catalog No. P19052G



Specifications

Specifications

Piroxicam
≥98.0% (HPLC,T)
MFCD00057317
4-Hydroxy-2-methyl-3-(2-pyridylcarbamoyl)-2H-1,2-benzothiazine 1,1-Dioxide, 4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
CN1C(=C(C2=CC=CC=C2S1(=O)=O)O)C(=O)NC3=CC=CC=N3
331.346
CHEBI:8249
Crystal-Powder at 20°C
201°C
36322-90-4
C15H13N3O4S
2811
QYSPLQLAKJAUJT-UHFFFAOYSA-N
4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1$l^{6},2-benzothiazine-3-carboxamide
54676228
331.35
White-Yellow
2g

Chemical Identifiers

36322-90-4
331.346
QYSPLQLAKJAUJT-UHFFFAOYSA-N
54676228
4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1$l^{6},2-benzothiazine-3-carboxamide
C15H13N3O4S
MFCD00057317
4-Hydroxy-2-methyl-3-(2-pyridylcarbamoyl)-2H-1,2-benzothiazine 1,1-Dioxide, 4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
CHEBI:8249
CN1C(=C(C2=CC=CC=C2S1(=O)=O)O)C(=O)NC3=CC=CC=N3
Safety and Handling

Safety and Handling

RTECSNumber : DL0705000

TSCA : No

Recommended Storage : Refrigerator

Documents