Learn More
Piroxicam, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6323909
Description
Piroxicam is an effective and potent inhibitor of prostaglandin synthesis and a Cox-1 and Cox-2 inhibitor. Studies show that Piroxicam blocks release of platelet ADP, and inhibits Cox-1 more potently than Cox-2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPiroxicam is an effective and potent inhibitor of prostaglandin synthesis and a Cox-1 and Cox-2 inhibitor. Studies show that Piroxicam blocks release of platelet ADP, and inhibits Cox-1 more potently than Cox-2.
Solubility
Soluble in water, ethanol, chloroform, ethyl acetate.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Piroxicam | |
Yellow | |
MFCD00057317 | |
UN2811 | |
piroxicam, feldene, piroxicamum, pyroxycam, roxicam, piroftal, baxo, artroxicam, bruxicam, caliment | |
QYSPLQLAKJAUJT-UHFFFAOYSA-N | |
4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1$l^{6},2-benzothiazine-3-carboxamide | |
54676228 | |
331.35 |
36322-90-4 | |
C15H13N3O4S | |
10 g | |
14,7506 | |
Soluble in water,ethanol,chloroform,ethyl acetate. | |
CN1C(=C(C2=CC=CC=C2S1(=O)=O)O)C(=O)NC3=CC=CC=N3 | |
331.346 | |
CHEBI:8249 | |
Powder |
Chemical Identifiers
36322-90-4 | |
331.346 | |
QYSPLQLAKJAUJT-UHFFFAOYSA-N | |
54676228 | |
4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1$l^{6},2-benzothiazine-3-carboxamide |
C15H13N3O4S | |
MFCD00057317 | |
piroxicam, feldene, piroxicamum, pyroxycam, roxicam, piroftal, baxo, artroxicam, bruxicam, caliment | |
CHEBI:8249 | |
CN1C(=C(C2=CC=CC=C2S1(=O)=O)O)C(=O)NC3=CC=CC=N3 |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 252-974-3
RTECSNumber : DL0705000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only