Please login to your online account to display your discounted pricing

Pirlindole mesylate, Tocris


MAO-A inhibitor

Quantity 10mg

Manufacturer: r & d systems  072410

 View more versions of this product

Catalog No. 072410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 322.42
Chemical Name or Material Pirlindole mesylate
Assay Percent Range >98%
Molecular Formula C15H18N2·CH3SO3H
CAS 207572-66-5
Synonym 2,3,3a,4,5,6-Hexahydro-8-methyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Pirlindole mesylate
Assay Percent Range >98%
Molecular Formula C15H18N2·CH3SO3H
Formula Weight 322.42
Synonym 2,3,3a,4,5,6-Hexahydro-8-methyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate
Accessories

Store at Room Temperature