Please login to your online account to display your discounted pricing

Pirlindole mesylate, Tocris

MAO-A inhibitor

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Pirlindole mesylate
Assay Percent Range >98%
Molecular Formula C15H18N2·CH3SO3H
Formula Weight 322.42
Synonym 2,3,3a,4,5,6-Hexahydro-8-methyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

072450

 
r & d systems
072450
50mg Each for $682.67

072410

 
r & d systems
072410
10mg Each for $164.61
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 322.42
Chemical Name or Material Pirlindole mesylate
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
Synonym 2,3,3a,4,5,6-Hexahydro-8-methyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate
Molecular Formula C15H18N2·CH3SO3H
CAS 207572-66-5
Safety and Handling

Store at Room Temperature