missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Piribedil, TCI America™

Manufacturer:  TCI America P20541G

 View more versions of this product

Catalog No. P20541G



Specifications

Specifications

Piribedil
98°C
3605-01-4
MFCD00868264
DEZKFCIOSKCCHK-UHFFFAOYSA-N
2-[4-(1,3-benzodioxol-5-ylmethyl)piperazin-1-yl]pyrimidine
4850
Crystal-Powder at 20°C
White
1g
C17H22N4O5S
2-[4-(1,3-Benzodioxol-5-ylmethyl)-1-piperazinyl]pyrimidine
C1CN(CCN1CC2=CC3=C(C=C2)OCO3)C4=NC=CC=N4
394.45
298.35
≥98.0% (GC,T)

Chemical Identifiers

3605-01-4
394.45
DEZKFCIOSKCCHK-UHFFFAOYSA-N
4850
C1CN(CCN1CC2=CC3=C(C=C2)OCO3)C4=NC=CC=N4
C17H22N4O5S
MFCD00868264
2-[4-(1,3-Benzodioxol-5-ylmethyl)-1-piperazinyl]pyrimidine
2-[4-(1,3-benzodioxol-5-ylmethyl)piperazin-1-yl]pyrimidine
Safety and Handling

Safety and Handling

RTECSNumber : UV9704000

TSCA : No

Documents